missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine EP Impurity G (Amlodipine Related Compound C), Mikromol™

Product Code. 30052373
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30052373

Brand: Mikromol MM0383.070025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Amlodipine EP Impurity G (Amlodipine Related Compound C)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material dimethyl 4-(2-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CAS 43067-01-2
Analyte or Component Names dimethyl 4-(2-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Shipping Condition Room Temperature
Molecular Formula C17 H18 Cl N O4
Certifications/Compliance ISO 17025
Synonym Dimethyl 4-(2-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Besilate Imp. G (EP), Amlodipine Imp. G (EP), Amlodipine USP Related Compound C, Amlodipine USP RC C, Dimethyl 4-(2-Chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, Amlodipine Related Compound C, Amlodipine Besilate Impurity G, Amlodipine Impurity G
InChI Formula InChI=1S/C17H18ClNO4/c1-9-13(16(20)22-3)15(11-7-5-6-8-12(11)18)14(10(2)19-9)17(21)23-4/h5-8,15,19H,1-4H3
SMILES COC(=O)C1=C(C)NC(=C(C1c2ccccc2Cl)C(=O)OC)C
IUPAC Name dimethyl 4-(2-chlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Weight (g/mol) 335.78
Quantity 25 mg
Formula Weight 335.0924 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.