missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminopeptidase N/CD13 Antibody (6E9), Novus Biologicals™
Rabbit Monoclonal Antibody
343.00 EUR - 533.00 EUR
Specifications
| Antigen | Aminopeptidase N/CD13 |
|---|---|
| Clone | 6E9 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043776
|
Novus Biologicals
NBP3-26507-50ul |
50 μL |
343.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043566
|
Novus Biologicals
NBP3-26507-100ul |
100 μL |
533.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aminopeptidase N/CD13 Monoclonal antibody specifically detects Aminopeptidase N/CD13 in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| Aminopeptidase N/CD13 | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, AP-M, AP-N, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPNhAPN | |
| A synthesized peptide derived from Human Aminopeptidase N/CD13 [UniProt P15144] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 6E9 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cellular Markers, Hematopoietic Stem Cell Markers, Immunology, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Stem Cell Markers | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 290 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title