Learn More
MP Biomedicals Amino Acid Diet, Synthetic, Powder
Product Code. p-7220720 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 kg
20 kg
50 kg
Unit Size:
10kg
20kg
50kg
Product Code. 11437341
Brand: MP Biomedicals 0296036110
Diet rich in Amino Acids. Can be added into agar gel to increase palatability.
  • Diet rich in amino acids
  • Can be added into agar gel to increase palatability
  • Can be used for research
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Product Type Animal Diet
Disposable Single-use
Format Powder
Quantity 10 kg
Sterility Non-sterile
Storage Requirements Refrigerated
Length (English) 1 ft.
Length (Metric) 0.3048 m
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.