missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Amidine Latex Beads, 4% w/v, 0.04μm

Product Code. 11594057
Change view
Click to view available options
Quantity:
15 mL
Unit Size:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11594057 15 mL 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11594057 Supplier Molecular Probes™ Supplier No. A37310

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Amidine Latex Beads, 4% w/v, 0.04 μm

Microspheres (also called latex beads or latex particles) are spherical particles in the colloidal size range that are formed from an amorphous polymer such as polystyrene. Because of the way that the polystyrene chains arrange themselves in the bead, the surface is very hydrophobic in character, making these ideal materials for the adsorption of materials such as proteins.

This positively-charged hydrophobic amidine latex is particularly suited for the preparation of latex intermediates. The only surface functional group present on the particle is amidine, which is not sensitive to the aggregating effects of polyvalent cations. These particles are sensitive to negatively charged contaminants or multivalent anions. The amidine latexes should be used in low to neutral pH environments. The amidine group is a base and with a pK ∼10-11.

Microspheres Are Available in Many Sizes and Surface Functionalities
  • 25 sizes to choose from—0.02 microns to 16 microns
  • 20 different surface—for ready conjugation to virtually any material
  • Surfactant free—ultraclean
  • Various quantities—from milliliter to 500-liter amounts

    Key Applications of Microspheres
  • Instrument calibration (flow cytometry, microscopy, HTS, HCS)
  • Flow testing (microfluidics, blood flow, water flow, and air flow)
  • Cell biology tracers (cell differentiation and cell tracing)
  • Immunoassays (agglutination tests, ELISA, particle capture, and contrast reagents)

    Get More Information
    Read a technical overview of microspheres and find out about high activity beads, super active beads, specialty beads, and review bead-coupling protocols on the IDC Surfactant-free Latex Beads application page.

    We’ll Make a Custom Microsphere Product for You
    If you don’t see the microsphere you’re looking for in our stocked list, we’ll make a custom microsphere for you. Our custom conjugation service is efficient and confidential, and we guarantee the quality of our work. We are ISO 9001:2000 certified.

    For Research Use Only. Not intended for animal or human therapeutic or diagnostic use.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Concentration 0.04w/v
    Content And Storage 2°C to 8°C
    Description Amidine Latex Beads, 15mL, 4% w/v Concentration, 0.04μm Particle Size, White, Suspension Form, De-ionized Water Medium, High Activity Latex Beads, Room Temperature, 2 to 8°C Storage Temperature, Ideal Materials for the Adsorption of Materials Such
    Color White
    For Use With (Application) Instrument Calibration (Flow Cytometry, Microscopy, HTS, HCS), Flow Testing (Microfluidics, Blood Flow, Water Flow, and Air Flow), Cell Biology Tracers (Cell Differentiation and Cell Tracing), Immunoassays (Agglutination Tests, ELISA, Particle Capture, and Contrast Reagents)
    Material Polystyrene
    Format Suspension
    Packaging 1 Each
    Parking Area 472 A2/C(NH)NH2 (Amidine Group)
    Particle Density 1.055g/cm3 at 20°C
    Quantity 15 mL
    Refractive Index 1.591 at 20°C
    Surface Modification Amidine
    Volume (Metric) 15 mL
    Diameter (Metric) Mean 0.04 μm
    Diameter (Metric) 0.04 μm
    Show More Show Less

    For Research Use Only. Not for use in diagnostic procedures.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.