Learn More
Cytiva Amersham™ Hybond™ -XL Membranes
Product Code. p-3675951 Shop All Cytiva Products
Click to view available options
Quantity:
10 Sheets
Length (Metric):
20 cm
Width (Metric):
20 cm
Format:
Sheet
Unit Size:
Pack of 10
Product Code. 12120960
Brand: Cytiva RPN2020S
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use for optimal signal-to-noise ratios when using radioactively labeled probes. Cytiva Life Sciences™ Amersham™ Hybond™ -XL Membranes are novel, charged nylon membranes that have a greater concentration and a more even distribution of positive charge than other charged nylon membranes.
  • Greater concentration and more even distribution of positive charge than other charged nylon membranes
  • Binding capacity for nucleic acids up to 600μg/cm2
  • Efficient binding of small fragments
  • Effective probe removal
  • Suitable for reprobing
  • Discs are also available
TRUSTED_SUSTAINABILITY
Specifications
Length (Metric) 20 cm
Width (Metric) 20 cm
For Use With (Equipment) Radioactive probes
Binding Capacity Up to 600μg/cm2
Quantity 10 Sheets
Format Sheet
Material Nylon, Highly Positively Charged