missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ameloblastin Polyclonal Antibody
GREENER_CHOICE

Product Code. 17151294 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17151294 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17151294 Supplier Invitrogen™ Supplier No. PA5113434

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ameloblastin Polyclonal Antibody for Western Blot, ELISA

This gene encodes the nonamelogenin enamel matrix protein ameloblastin. The encoded protein may be important in enamel matrix formation and mineralization. This gene is located in the calcium-binding phosphoprotein gene cluster on chromosome 4. Mutations in this gene may be associated with dentinogenesis imperfect and autosomal dominant amylogenesis imperfect.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ameloblastin
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene AMBN
Gene Accession No. Q62840, Q9NP70
Gene Alias AI1F; AMBN; Amelin; ameloblastin; ameloblastin (enamel matrix protein); ameloblastin precursor; enamel matrix protein
Gene Symbols AMBN
Host Species Rabbit
Immunogen Recombinant Human Ameloblastin protein (264-387AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25376, 258
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.