missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambroxol (L)-Lactamide tert-Butyl Carbamate, TRC
Specifications
Specifications
| Chemical Name or Material | Ambroxol (L)-Lactamide tert-Butyl Carbamate |
| Analyte or Component Names | Ambroxol (L)-Lactamide tert-Butyl Carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H30 Br2 N2 O5 |
| Synonym | tert-Butyl (3,5-dibromo-2-((S)-2-hydroxypropanamido)benzyl)((1r,4S)-4-hydroxycyclohexyl)carbamate |
| InChI Formula | InChI=1S/C21H30Br2N2O5/c1-12(26)19(28)24-18-13(9-14(22)10-17(18)23)11-25(20(29)30-21(2,3)4)15-5-7-16(27)8-6-15/h9-10,12,15-16,26-27H,5-8,11H2,1-4H3,(H,24,28)/t12-,15-,16-/m0/s1 |
| Solution Type | Neat |
| SMILES | C[C@H](O)C(=O)Nc1c(Br)cc(Br)cc1CN([C@@H]2CC[C@@H](O)CC2)C(=O)OC(C)(C)C |
| IUPAC Name | tert-butyl N-[[3,5-dibromo-2-[[(2S)-2-hydroxypropanoyl]amino]phenyl]methyl]-N-(4-hydroxycyclohexyl)carbamate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?