Learn More
Invitrogen™ PSI-CLONE™ 96-Well PCR Purification Plates
Description
Applied Biosystems PSI-Clone™ PCR 96-Well plates are for the rapid purification of PCR reaction products using either vacuum- or centrifugation-based protocols. Both protocols are fast and result in high-yield, high-purity DNA ideal for molecular biology experiments.
DNA and RNA Purification and Analysis, DNA Extraction, PCR Product Clean-Up
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Format | 96-well Plate |
| Quantity | 4 Plates |