Learn More
Invitrogen™ Ambion™ MEGAshortscript™ T7 Transcription Kit
Specifications
Specifications
| Content And Storage | T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-RNA 18S, TURBO DNase, Ammonium Acetate Stop Solution, and Gel Loading Buffer II are all stored at -20°C. Nuclease-free Water may be stored at any temperature. |
| For Use With (Application) | Gene Expression Analysis and Genotyping, in vitro Transcription |
| Includes | T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-RNA 18S, TURBO DNase, Ammonium Acetate Stop Solution, Gel Loading Buffer II, and Nuclease-free Water |
| No. of Reactions | 25 |
| Product Type | Kit with manual |
| Quantity | 25 Reactions |