Learn More
Invitrogen™ MagMAX™ Lysis/Binding Solution Concentrate
Product Code. 10311425
Click to view available options
Quantity:
100 mL
Unit Size:
Each
Product Code. 10311425
Brand: Invitrogen™ AM8500
Invitrogen MagMAX Lysis/Binding Solution Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately.
Invitrogen MagMAX Lysis/Binding Solution Concentrate is a component of all the MagMAX Isolation Kits that may be purchased separately. Supplied in one 100 mL bottle.
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Buffer Lysis Buffer, Binding Buffer
Content And Storage Store MagMAX™ Lysis/Binding Solution Concentrate at room temperature.
Product Line Ambion, MagMAX
Quantity 100 mL
Shipping Condition Room Temperature