Learn More
Fisherbrand™ Amber Borosilicate Glass Class A Volumetric Flask with Stopper
Product Code. p-8002008 Shop All Fisherbrand Products
Click to view available options
Capacity (Metric):
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
Closure Size:
10/19
12/21
14/23
19/24
24/29
Tolerance:
±0.01 mL
±0.04 mL
±0.06 mL
±0.15 mL
±0.25 mL
±0.4 mL
Unit Size:
Pack of 2
Pack of 5
Product Code. 11359443
Only null left
Add to basket
Add to basket
Amber Borosilicate Glass Volumetric Flask includes interchangeable polyethylene stopper.
With one mark graduation and interchangeable polyethylene stopper. The details on each flask include confirmation of compliance to International Standards as well as an individual re-order code. The Fisherbrand 'Batch Certificate of Compliance' is downloadable for this item from the 'Documentation' tab
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 1042, DIN 12664
Closure Material Polyethylene
Color Amber
Capacity (Metric) 1000 mL
Material Borosilicate Glass
Tolerance ±0.4 mL
Graduated Yes
Class A
Closure Size 24/29