Learn More
AM095 free acid, MedChemExpress
Product Code. 16406978
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
Product Code. 16406978
Brand: Medchem Express HY16040
MedChemExpress AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.
AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.
Specifications
Chemical Name or Material AM095 free acid
CAS 1228690-36-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C27H24N2O5
Quantity 10 mg
Solubility Information DMSO : 67.3 mg/mL (147.43 mM; Need ultrasonic and warming)
SMILES O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4)C)=O)C(C)=NO3)C=C2)C=C1
Molecular Weight (g/mol) 456.49
Formula Weight 456.49
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation