Learn More
VACUUBRAND™ Aluminum Vacuum Tubing With Small Flanges KF
Product Code. 10779152
Click to view available options
Tubing Size:
80mm
130mm
Unit Size:
Each
Product Code. 10779152
Brand: VACUUBRAND™ 20669014
Only null left
Add to basket
Add to basket
Providing durable connection solutions for the laboratory.
  • Manufactured from aluminum
  • Available in a range of sizes and lengths
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Diameter (Metric) Outer 16 mm
Overall Length (Metric) 80 mm
Packaging Quantity 1 Each
Tubing Size 80mm
Includes Small flanges
Product Type Vacuum Tubing