Learn More
Cole-Parmer™ Aluminum Scoop
Shop All Cole Parmer Products
Click to view available options
Capacity (Metric):
350 mL
1100 mL
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 1100 mL |
| Product Type | Scoop |
| Length (English) | 9 in. |
| Material | Aluminum |
| Handle Type | Ribbed |