Learn More
Cole-Parmer™ Essentials Round Support Plate
Product Code. 11705359 Shop All Cole Parmer Products
Click to view available options
Material:
Aluminum
Unit Size:
Each
Product Code. 11705359
Brand: Cole-Parmer™ Essentials 0802716
Only null left
Add to basket
Add to basket
Provide support for your specific laboratory equipment with this Round Support Plate.
  • Use to support your labware items from lab frames or support stands.
  • The round plate design features a raised rim to prevent your apparatus from falling.
  • Requires an additional connector in order to attach clamps to frames or other apparatus.
  • The rectangular plates include an integral connector.
Specifications
Material Aluminum
Diameter (English) Outer 152.4 mm
Diameter (English) Rod 12.7 mm
Diameter (Metric) Outer 6 in.
Diameter (Metric) Rod 0.5 in.
Product Type Specialty Supports
Shape Rectangular
For Use With (Application) For Specific Laboratory Equipment
Quantity 1 Ea.