Learn More
Bochem™ Aluminum Rods
Click to view available options
Length (Metric) Rod:
500 mm
600 mm
750 mm
1000 mm
Unit Size:
Each
Description
- Aluminum
- Without thread
- Sizes vary
- For stand base
- 12 D/O
Specifications
Specifications
| Material | Aluminum |
| Length (Metric) Rod | 1000 mm |
| Diameter (Metric) Rod | 12 dia. |
| For Use With (Application) | Stand Bases |