missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum, Reference Material, AA 6061, A96061, ARMI∣MBH
Click to view available options
Form:
Disc
Pieces
Unit Size:
50g
Each
Description
Aluminum, Reference Material, AA 6061, A96061
Specifications
Specifications
| Chemical Name or Material | Chromium, Copper, Gallium, Iron, Magnesium, Manganese, Nickel, Silicon, Titanium, Vanadium, Zinc |
| Diameter (Metric) Outside | 38 mm |
| Form | Disc |
| Thickness (Metric) | 6 mm |
| Concentration or Composition (by Analyte or Components) | Zinc: 0.002%, Nickel: 0.003%, Vanadium: 0.014%, Gallium: 0.015%, Titanium: 0.028%, Manganese: 0.068%, Chromium: 0.198%, Copper: 0.266%, Iron: 0.269%, Silicon: 0.605%, Magnesium: 0.886% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction