Learn More
Brady™ Aluminum with Polyester Overlaminate Warning Safety Signs
Product Code. 11886371
Click to view available options
Width (Metric):
100 mm
200 mm
Displayed Text:
Danger Zone Warning Pictogram - !
Unit Size:
Each
Product Code. 11886371
Brand: Brady™ 255216
Specifications
Certifications/Compliance ISO 7010, EC 95/98 Standard
Alternate Part Number PIC 308-TRI 200-B7525
Material Aluminum, Polyester
Quantity 1 Ea.
Description Aluminum with Polyester Overlaminate
For Use With (Application) Used for indoor and outdoor safety signs
Thickness 0.271 mm
Width (Metric) 200 mm
Temperature Range (Metric) -40°C to 140°C
Shape Triangular
Displayed Text Danger Zone Warning Pictogram - !
Product Type Warning Safety Sign
Color Black on Yellow