Learn More
Bochem™ Aluminum Laboratory Pot With Lid
30.60 EUR - 41.50 EUR
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10772352
|
Bochem™
8566 |
1.5 L |
30.60 EUR
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
|||||
10044920
|
Bochem™
8567 |
3 L |
35.05 EUR
Each |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||||
10054920
|
Bochem™
8568 |
5.2 L |
41.50 EUR
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Specifications
Laboratory Pot | |
Aluminum | |
Lid |