Learn More
Fisherbrand™ Aluminum Labjacks
Shop All Fisherbrand Products
Click to view available options
Height (Metric):
128 to 470 mm
36 to 78 mm
43 to 140 mm
57 to 290 mm
60 to 290 mm
Length (Metric) Base:
50 mm
100 mm
150 mm
200 mm
320 mm
Width (Metric) Base:
40 mm
100 mm
150 mm
200 mm
260 mm
Unit Size:
Each
Specifications
Specifications
| Material | Aluminum |
| Height (Metric) | 36 to 78 mm |
| Length (Metric) Base | 50 mm |
| Width (Metric) Base | 40 mm |
| Includes | With stainless steel operating screw |
| Product Type | Lab Jack |
| Quantity | 1 Ea. |