Learn More
KGW Isotherm™ Dish-Shaped Dewar Flasks
Click to view available options
Capacity (Metric):
260 mL
400 mL
800 mL
5300 mL
Unit Size:
Each
Description
- Easy handling
- Suitable on magnetic stirrer
- Full silvered
- Made of borosilicate glass 3.3
- Polyethylene lid (not included)
Specifications
Specifications
| Diameter (Metric) Outer | 120mm |
| For Use With (Application) | Cooling of small samples |
| Height (Metric) Inner | 65mm |
| Height (Metric) | 110mm |
| Capacity (Metric) | 260 mL |
| Diameter (Metric) Inner | 100 mm |
| Material | Borosilicate Glass |