Learn More
Texwipe™ AlphaLite™ II Cleanroom Floor Wipes
Product Code. 11565395 Shop All Texwipe Products
Change view
Click to view available options
Dimensions (L x W):
600 x 250 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
11565395 600 x 250 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11565395 Supplier Texwipe™ Supplier No. TX3424
Only null left
Add to basket
Add to basket
Cut-edge polyester single ply wipe that is ideal for floor cleaning and for applying cleaning solutions and disinfectants on floors
  • Large size for improved application on floor surfaces
  • Size is compatible with most mopping systems
  • Improved contamination control
  • Suitable for use in Class 100, ISO 5 and Class 1000, ISO 6 environments
Specifications
Dimensions (L x W) 600 x 250 mm
For Use With (Application) Floor cleaning and for applying cleaning solutions and disinfectants on floors
Packaging Quantity 10 Wipes/Bag, 34 Bags/Cs.