missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Monothioglycerol, 99.1%, For GC analysis, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 mL
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
25mL
50mL
Specifications
Specifications
| Color | Colorless |
| Quantity | 25 mL |
| Formula Weight | 108.2 |
| Percent Purity | 0.991 |
| Physical Form | Liquid |
| Chemical Name or Material | α-Monothioglycerol, For GC analysis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction