missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Fructoselysine Dihydrochloride, TRC

Product Code. 30133090
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
50 mg
Unit Size:
2.5mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133090 50 mg 50mg
30138633 2.5 mg 2.5mg
30007469 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133090 Supplier Toronto Research Chemicals Supplier No. TRCF79570050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96192-34-6
Molecular Formula C12H24N2O7 . 2(HCl)
Molecular Weight (g/mol) 308.3323646
Synonym (S)-1-[(5-Amino-1-carboxypentyl)amino]-1-deoxy-D-fructose Dihydrochloride, Fructoselysine Dihydrochloride, Fructosyllysine Dihydrochloride, Nα-(1-Deoxy-D-fructos-1-yl)-L-lysine Dihydrochloride;
SMILES OC1(CN[C@H](C(O)=O)CCCCN)[C@@H](O)[C@H](O)[C@H](O)CO1>>OC[C@@H]2[C@@H](O)[C@H](O)C(O)(CN[C@H](C(O)=O)CCCCN)O2

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H24N2O7/c13-4-2-1-3-7(11(18)19)14-6-12(20)10(17)9(16)8(15)5-21-12/h7-10,14-17,20H,1-6,13H2,(H,18,19)/t7-,8+,9+,10-,12?/m0/s1
Chemical Name or Material alpha-Fructoselysine Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.