missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Fetoprotein/AFP Rabbit anti-Human, Clone: AFP/7007R, Novus Biologicals™
Rabbit Monoclonal Antibody
292.00 EUR - 528.00 EUR
Specifications
| Antigen | alpha-Fetoprotein/AFP |
|---|---|
| Clone | AFP/7007R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18341266
|
Novus Biologicals
NBP3-13870-20UG |
20 μg |
292.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18394054
|
Novus Biologicals
NBP3-13870-100UG |
100 μg |
528.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
alpha-Fetoprotein/AFP Monoclonal antibody specifically detects alpha-Fetoprotein/AFP in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| alpha-Fetoprotein/AFP | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cancer Stem Cells, Developmental Biology, GPCR, Stem Cell Markers, Tumor Biomarkers | |
| 10 mM PBS with 0.05% BSA | |
| Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
| Recombinant human full-length alpha-Fetoprotein/AFP protein (Uniprot: P02771 ) | |
| Primary | |
| Store at 4°C. |
| AFP/7007R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P02771 | |
| 174 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title