missing translation for 'onlineSavingsMsg'
Learn More

Alpha-Chloro-p-methyltoluene, TRC

Product Code. 30138326
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
Unit Size:
100g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138326 100 g 100g
30007782 25 g 25g
30003525 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138326 Supplier Toronto Research Chemicals Supplier No. TRCC369770100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-82-5
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
Synonym alpha-Chloro-p-xylene, (4-Methylphenyl)methyl Chloride, 1-(Chloromethyl)-4-methylbenzene, 1-Methyl-4-chloromethylbenzene, 4-(Chloromethyl)toluene, 4-Methylbenzyl Chloride, NSC 46590, p-(Chloromethyl)toluene, p-Methylbenzyl Chloride, p-Tolylmethyl Chloride, p-Xylyl Chloride, p-Xylyl-alpha-chloride, alpha-Chloro-p-methyltoluene, alpha-Chloro-p-xylene
IUPAC Name 1-(chloromethyl)-4-methylbenzene
SMILES Cc1ccc(CCl)cc1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C8H9Cl/c1-7-2-4-8(6-9)5-3-7/h2-5H,6H2,1H3
Formula Weight 140.0393
Chemical Name or Material alpha-Chloro-p-methyltoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.