missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha-1-Antichymotrypsin Mouse, Clone: AACT/1451, enQuireBio™
Mouse Monoclonal Antibody
199.00 EUR - 478.00 EUR
Specifications
| Antigen | Alpha-1-Antichymotrypsin |
|---|---|
| Clone | AACT/1451 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15949807
|
EnquireBio
12-MSM1-P0 |
20 μg | |||||||
|
15959807
|
EnquireBio
12-MSM1-P1 |
100 μg |
554.40 EUR 478.00 EUR / 100µg Save 76.40 EUR 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Alpha-1-Antichymotrypsin | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 12 | |
| Recombinant human Antichymotrypsin (AACT) protein fragment (aa49-187) | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 65-76kDa |
| AACT/1451 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_001076 | |
| SERPINA3 | |
| IgG1 | |
| Protein A/G Purified | |
| Cytoplasmic | |
| HeLa Cells. Tonsil, Pancreas or Histiocytoma. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title