missing translation for 'onlineSavingsMsg'
Learn More

Allyl methyl carbonate, 98%

Product Code. 10055253 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055253 25 mL Glass bottle 25mL
10350574 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10055253 Supplier Thermo Scientific Acros Supplier No. 389750250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35466-83-2 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35466-83-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00134545
InChI Key YHLVIDQQTOMBGN-UHFFFAOYSA-N
Synonym allyl methyl carbonate, methyl prop-2-en-1-yl carbonate, carbonic acid allyl methyl ester, isobutylene carbonate, methyl allyl carbonate, acmc-1agon, allyl methyl carbonate #, prop-2-enyl methoxyformate, yhlvidqqtombgn-uhfffaoysa
PubChem CID 534788
IUPAC Name methyl prop-2-enyl carbonate
SMILES COC(=O)OCC=C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0220g/mL
Boiling Point 131.0°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2=CHCH2OCO2CH3
Refractive Index 1.4050 to 1.4070 (20°C, 589nm)
Quantity 25 mL
Specific Gravity 1.022
Formula Weight 116.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allyl methyl carbonate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Allyl methyl carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.