missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alloy Steel, Ni/Cr/Mo, AISI 4340, G43400, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Alloy Steel, Ni/Cr/Mo, AISI 4340, G43400
Specifications
Specifications
| Chemical Name or Material | Aluminium, Antimony, Arsenic, Boron, Calcium, Carbon, Chromium, Cobalt, Copper, Iron, Lead, Manganese, Molybdenum, Nickel, Niobium, Nitrogen, Phosphorus, Silicon, Sulfur, Tin, Titanium, Tungsten, Vanadium |
| Diameter (Metric) Outside | 38 mm |
| Form | Disc |
| Thickness (Metric) | 19 mm |
| Concentration or Composition (by Analyte or Components) | Calcium: 21 ppm, Antimony: 12 ppm, Boron: 4 ppm, Lead: 0.29 ppm, Titanium: 0.001%, Niobium: 0.003%, Vanadium: 0.0047%, Arsenic: 0.0055%, Nitrogen: 0.007%, Phosphorus: 0.0079%, Tin: 0.0083%, Tungsten: 0.011%, Aluminium: 0.022%, Sulfur: 0.0273%, Cobalt: 0.093%, Copper: 0.174%, Molybdenum: 0.242%, Silicon: 0.244%, Carbon: 0.382%, Manganese: 0.739%, Chromium: 0.787%, Nickel: 1.75% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction