missing translation for 'onlineSavingsMsg'
Learn More

Allicin, MedChemExpress

Product Code. 16414817
Encompass_Preferred
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 16414817

Brand: Medchem Express HYN0315

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Allicin (diallyl thiosulfinate) is isolated from garlic including Diallyl monosulfide, Diallyl disulfide, Diallyl trisulfide, Diallyl tetrasulfide, and Methyl allyl disulphide etc. They accounts for 98% of the extract. Allicin (diallyl thiosulfinate) has highly potent antimicrobial activity, and inhibits growth of a variety of microorganisms, among them antibiotic-resistant strains.

Allicin (diallyl thiosulfinate) is isolated from garlic including Diallyl monosulfide, Diallyl disulfide, Diallyl trisulfide, Diallyl tetrasulfide, and Methyl allyl disulphide etc. They accounts for 98% of the extract. Allicin (diallyl thiosulfinate) has highly potent antimicrobial activity, and inhibits growth of a variety of microorganisms, among them antibiotic-resistant strains.

Specifications

Chemical Name or Material Allicin
Physical Form Liquid
CAS 539-86-6
Color Colorless
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C6H10OS2
For Use With (Application) COVID-19-immunoregulation
Synonym Diallyl thiosulfinate
Solubility Information DMSO : 130 mg/mL (801.13 mM; Need ultrasonic) ∣Ethanol : 70 mg/mL (431.38 mM; Need ultrasonic)
SMILES C=CCS(SCC=C)=O
Molecular Weight (g/mol) 162.27
Formula Weight 162.27
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.