Learn More
S Murray™ Samco™ All Glass Luer-Tip Syringe
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
20 mL
Unit Size:
Each
Description
Use these syringes for a variety of research applications. S Murray™ All Glass Luer-Tip Syringes are graduated and feature glass luer fittings.
- Syringe with glass luer fitting
- Fully interchangeable
Specifications
Specifications
| Certifications/Compliance | CE |
| Material | Glass |
| Product Type | All Glass Luer-tip Syringe |
| Quantity | 1/Ea. |
| Volume (Metric) | 1 mL |
| Connection Type | Luer Tip |
| Graduated | Yes |