missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Alkyne-cRGD

Click to view available options
Quantity:
2 mg
Unit Size:
2mg
Description
Alkyne-cRGD is an integrin ligand for degrader development and targeted delivery. It contains an alkyne handle that enables the attachment of additional groups via copper-catalyzed azide-alkyne cycloaddition click reactions.
Specifications
Specifications
| Purity | 95% |
| CAS | 2665674-77-9 |
| Content And Storage | Store at -20°C |
| Molecular Weight (g/mol) | M.W. (Observed): 713.78 |
| Quantity | 2 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction