Learn More
Thermo Scientific™ Alkaline Saline Peptone Water (ISO) (Dehydrated)
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
missing translation for 'unitSize'
500g
Description
Enrich and recover Vibrio species from food and water samples with Thermo Scientific™ Oxoid™ Alkaline Saline Peptone Water (ISO) (Dehydrated), while adhering to ISO/TS 21872-1&2:2007. This medium is selective by exploiting the ability of Vibrio species to tolerate alkaline conditions and high sodium chloride levels. The peptone content provides the conditions for growth and recovery of Vibrio species, supplying vitamins, minerals, and nitrogen in the form of amino acids.
Use Alkaline Saline Peptone Water (ISO) for selective recovery and enrichment for Vibrio species.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
- Creates strong alkaline and high sodium conditions to provide a selective environment for Vibrio species.
- Adheres to the ISO/TS 21872-1&2 2007 standards.
- Can be used as a non-selective enrichment broth for the cultivation of Aeromonas species.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Spécification
Spécification
| Content And Storage | 10°C to 30°C |
| Description | Alkaline Saline Peptone Water (ISO) (Dehydrated) |
| Form | Powder |
| Product Type | Pre-enrichment Medium |
| Quantity | 500 g |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu