missing translation for 'onlineSavingsMsg'
Learn More

Alizapride hydrochloride, MedChemExpress

Product Code. p-200096447
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
Product Code. Quantity unitSize
16464187 50 mg 50mg
16448036 100 mg 100mg
2 options
This item is not returnable. View return policy

Product Code. 16464187

Brand: Medchem Express HYA0125A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alizapride is a potent antiemetic, acting as a dopamine receptor antagonist. Alizapride also used in human digestive disorders.

Alizapride is a potent antiemetic, acting as a dopamine receptor antagonist. Alizapride also used in human digestive disorders.

Specifications

Chemical Name or Material Alizapride hydrochloride
CAS 59338-87-3
Color White
Purity Grade Notes Research
Molecular Formula C16H22ClN5O2
Quantity 50 mg
Solubility Information H2O : 100 mg/mL (284.23 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (94.73 mM; Need ultrasonic)
SMILES O=C(C1=C(OC)C=C(NN=N2)C2=C1)NCC3N(CC=C)CCC3.Cl[H]
Molecular Weight (g/mol) 351.83
Formula Weight 351.83
Percent Purity 98.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.