Learn More
Gibco™ AlgiMatrix™ Firming Buffer
Description
AlgiMatrix™ Firming Buffer is for use with the AlgiMatrix™ 3D Culture System to facilitate the formation of firm sponges in the 96 well and 24 well plate formats. This buffer allows the users to adjust the firmness of the sponge to meet specific cell and experimental requirements. Should be ordered as a separate component.
Specifications
Specifications
| Chemical Name or Material | Firming Buffer |
| For Use With (Equipment) | 3D Culture System to facilitate the formation of firm sponges in the 96 well and 24 well plate formats |
| Physical Form | Liquid |
| Product Line | Algimatrix |
| Quantity | 50 mL |
| Sterility | Sterile |
| Grade | Cell Culture Grade |