missing translation for 'onlineSavingsMsg'
Learn More

Alexamorelin Trifluoroacetic Acid Salt, TRC

Product Code. 30011474
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011474 5 mg 5mg
30006241 1 mg 1mg
30004980 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011474 Supplier Toronto Research Chemicals Supplier No. TRCA5249035MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C50 H63 N13 O7 . x(C2 H F3 O2)
Molecular Weight (g/mol) 1072.1
Synonym L-Alanyl-L-histidyl-2-methyl-D-tryptophyl-L-alanyl-L-tryptophyl-D-phenylalanyl-L-lysinamide Trifluoroacetic Acid Salt
IUPAC Name (2S)-6-amino-2-[[(2R)-2-[[(2S)-2-[[(2S)-2-[[(2R)-2-[[(2S)-2-[[(2S)-2-aminopropanoyl]amino]-3-(1H-imidazol-4-yl)propanoyl]amino]-3-(2-methyl-1H-indol-3-yl)propanoyl]amino]propanoyl]amino]-3-(1H-indol-3-yl)propanoyl]amino]-3-phenyl-propanoyl]amino]hexanamide;2,2,2-trifluoroacetic acid
SMILES C[C@H](N)C(=O)N[C@@H](Cc1c[nH]cn1)C(=O)N[C@H](Cc2c(C)[nH]c3ccccc23)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(=O)N[C@H](Cc6ccccc6)C(=O)N[C@@H](CCCCN)C(=O)N.OC(=O)C(F)(F)F

Specifications

Analyte or Component Names Alexamorelin Trifluoroacetic Acid Salt
Shipping Condition Room Temperature
InChI Formula InChI=1S/C50H63N13O7.C2HF3O2/c1-28(52)45(65)60-42(23-33-26-54-27-56-33)50(70)63-43(24-36-29(2)57-38-18-10-8-16-35(36)38)47(67)58-30(3)46(66)61-41(22-32-25-55-37-17-9-7-15-34(32)37)49(69)62-40(21-31-13-5-4-6-14-31)48(68)59-39(44(53)64)19-11-12-20-51;3-2(4,5)1(6)7/h4-10,13-18,25-28,30,39-43,55,57H,11-12,19-24,51-52H2,1-3H3,(H2,53,64)(H,54,56)(H,58,67)(H,59,68)(H,60,65)(H,61,66)(H,62,69)(H,63,70);(H,6,7)/t28-,30-,39-,40+,41-,42-,43+;/m0./s1
Solution Type Neat
Industry Type Forensics
Quantity 5 mg
Chemical Name or Material Alexamorelin Trifluoroacetic Acid Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.