Learn More
Molecular Probes™ Alexa Fluor™ 488 Signal-Amplification Kit for Mouse Antibodies
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Description
For sensitive detection of mouse primary antibodies, try our Alexa Fluor™ 488 Signal-Amplification Kit, which contains Alexa Fluor 488 rabbit anti—mouse IgG and Alexa Fluor 488 goat anti—rabbit IgG conjugates. This kit takes advantage of the intense, photostable fluorescence of Alexa Fluor 488 antibody conjugates and can be used to amplify the signals derived from mouse-derived antibodies. The kit contains a detailed protocol and sufficient reagents for ∼60 assays by flow cytometry or imaging. For added convenience, the kit is designed for use with filters and settings appropriate for fluorescein.
Specifications
Specifications
| Content And Storage | Store in refrigerator (2–8°C) and protect from light. |
| Label or Dye | Alexa Fluor™ 488 |
| Product Line | Alexa Fluor |
| Quantity | 1 kit |
| Shipping Condition | Room Temperature |