missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aldolase C Polyclonal Antibody
GREENER_CHOICE

Product Code. 10545890
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10545890 400 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10545890 Supplier Invitrogen™ Supplier No. PA512317

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse, non-human primate and rat based on sequence homology.

This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively. [provided by RefSeq, Jul 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldolase C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ALDOC
Gene Accession No. P05063, P09972
Gene Alias AI847350; ALDC; ALDCAA; Aldo3; ALDOC; Aldolase 3; aldolase C, fructose-biphosphate; aldolase C, fructose-bisphosphate; aldolase, fructose-bisphosphate C; AU040929; Brain-type aldolase; EC 4.1.2.13; F16dip7; fructoaldolase C; fructose-1,6-biphosphate triosephosphate lyase; fructose-bisphosphate aldolase C; RATALDCAA; Scrapie-responsive protein 2; Scrg2; Zebrin II
Gene Symbols ALDOC
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 300-333 amino acids from the C-terminal region of human ALDOC.
Purification Method Size-exclusion - Dialysis, Ammonium sulfate precipitation
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11676, 230
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.