missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies ALDHbr Assay Kit
Click to view available options
Quantity:
6 x 107 Cells
Unit Size:
1 set
Description
- The ALDHbr Assay Kit is optimized for the identification and quantitation of CD34+ cells that express high levels of the enzyme aldehyde dehydrogenase (ALDH) in human cord blood samples
- This kit contains both the CD34+ Cell Detection Kit and the ALDEFLUOR™ Kit
- The CD34+ Cell Detection Kit contains pre-selected fluorochrome-conjugated monoclonal antibodies that have been optimized for use with the ALDEFLUOR™ Kit to detect cells that express CD34 and have high ALDH activity by flow cytometry
- The CD34+ Cell Detection Kit contains anti-human antibodies APC CD34, PE CD45, and PE-Cyanine5 Glycophorin A/B, and 7-AAD viability dye
- A flow cytometer equipped with a 488 nm laser and a 635 nm laser for excitation and appropriate filters for detecting APC, PE, PE-Cyanine5 and 7-AAD fluorescence is required for use
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Flow Cytometry |
| Quantity | 6 x 107 Cells |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction