Learn More
ALDH1B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15560-1-AP-20UL
 View more versions of this product
Product Code. 16854794
  • 145.00 EUR / 20µL
Description
Aldehyde dehydrogenases (ALDHs) mediate NADP+-dependent oxidation of aldehydes into acids during detoxification of alcohol-derived acetaldehyde, lipid peroxidation and metabolism of corticosteroids, biogenic amines and neurotransmitters. Alcohol drinking habits and cardiovascular disease risk factors may be associated with ALDH gene variants. ALDH1B1 (Aldehyde dehydrogenase family 1 member B1), also known as ALDH5 or ALDHX (Aldehyde dehydrogenase X, mitochondrial), is a 517 amino acid mitochondrial protein that is expressed in the liver, testis and to a lesser extent in brain. ALDH1B1 belongs to the aldehyde dehydrogenase family and may play a major role in ethanol detoxification.
Specifications
Specifications
ALDH1B1
Polyclonal
Unconjugated
ALDH1B1
Aldehyde dehydrogenase 5, ALDH1B1, ALDH5, ALDHX
Rabbit
Antigen Affinity Chromatography
RUO
219, 298079, 72535
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P30837, Q66HF8, Q9CZS1
Aldh1b1
ALDH1B1 Fusion Protein Ag7144
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions