missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Albumin (Transport Protein) Mouse, Clone: ALB/2144, enQuireBio™
Mouse Monoclonal Antibody
199.00 EUR - 478.00 EUR
Specifications
| Antigen | Albumin (Transport Protein) |
|---|---|
| Clone | ALB/2144 |
| Concentration | 200μg/mL |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15969967
|
EnquireBio
213-MSM4-P0 |
20 μg | |||||||
|
15979967
|
EnquireBio
213-MSM4-P1 |
100 μg |
554.40 EUR 478.00 EUR / 100µg Save 76.40 EUR 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Albumin (Transport Protein) | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
| Recombinant full-length human ALB protein | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 66kDa |
| ALB/2144 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| 213 | |
| IgG1 κ | |
| Protein A/G purified from Bioreactor concentrate. | |
| Cell Surface and Cytoplasmic | |
| Human serum. Liver or Hepatocellular Carcinoma |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title