Learn More
Alachlor-d13 (2,6-diethylphenyl-d13), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Alachlor-d13 |
| CAS | 1015856-63-9 |
| Molecular Formula | C14 2H13 H7 Cl N O2 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C14H20ClNO2/c1-4-11-7-6-8-12(5-2)14(11)16(10-18-3)13(17)9-15/h6-8 H,4-5,9-10H2,1-3H3/i1D3,2D3,4D2,5D2,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c(c(N(COC)C(=O)CCl)c(c1[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-chloro-N-(methoxymethyl)-N-[3,4,5-trideuterio-2,6-bis(1,1,2,2,2-pentadeuterioethyl)phenyl]acetamide |
| Molecular Weight (g/mol) | 282.85 |
| Formula Weight | 282.1999 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |