Learn More
Al-24 Tube (both ends open);OD (mm), 30;ID (mm), 24
Product Code. 11334117
Click to view available options
Quantity:
100 mm
200 mm
300 mm
600 mm
800 mm
1000 mm
Unit Size:
1000mm
100mm
200mm
300mm
600mm
800mm
Product Code. 11334117
Brand: Thermo Scientific Alfa Aesar 031606.EH
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-24 Tube
Name Note Both ends open
Quantity 600 mm
What is the maximum working temperature of Al-24 Tubes?
The maximum working temperature for the Al-24 Tube is 1950 degrees C.