Learn More
Al-23 Tube for Heating Coils, O.D.: 50mm, I.D.: 38mm, Pitch: 2.5, Thermo Scientific™
Product Code. 11323168
Change view
Click to view available options
Quantity:
100 mm
200 mm
300 mm
500 mm
Unit Size:
100mm
200mm
300mm
500mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11323168 100 mm 100mm
11393158 200 mm 200mm
11303168 300 mm 300mm
11313168 500 mm 500mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11323168 Supplier Thermo Scientific Alfa Aesar Supplier No. 033209.DM
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Al-23 Tube for Heating Coils
Quantity 100 mm