Learn More
Syngene™ Airclean 600 PCR Workstation
Shop All Syngene Products
Click to view available options
Width (English):
32 in.
48 in.
Unit Size:
Each
Description
- With UVTect microprocessor controller
- Provides Class 100 vertical laminar air flow and timed uv light for sterilization
- Includes HEPA filter, initial prefilter, fluorescent light, UV shelf with pipette holder, safety switch, and slip hatch
Specifications
Specifications
| Product Type | PCR Workstation |
| Certifications/Compliance | UL, CSA, and CE approved |
| Includes | UV Controller |
| Voltage | 220 V |
| Width (English) | 48 in. |
| Width (Metric) | 121.92 cm |