missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AIFM1 Monoclonal Antibody (OTI5D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811193
This item is not returnable.
View return policy
Description
Apoptosis Inducing Factor (AIF) causes chromatin condensation and DNA fragmentation. AIF was recently identified, and cloned. Apoptosis is characterized by several morphological nuclear changes including chromatin condensation and nuclear fragmentation. These changes are triggered by the activation of members of caspase family, caspase activated DNase, and several novel proteins. Like the critical molecules, cytochrome c and caspase-9, in apoptosis, AIF localizes in mitochondria. AIF translocates to the nucleus when apoptosis is induced and induces mitochondria to release the apoptogenic proteins cytochrome c and caspase-9. AIF induces chromatin condensation and large scale DNA fragmentation, which are the hallmarks of apoptosis, of the isolated nucleus and the nucleus in live cells by microinjection and apoptosis stimuli. AIF is highly conserved between human and mouse and widely expressed. Mutations in the AIF gene cause combined oxidative phosphorylation deficiency 6, which results in a severe mitochondrial encephalomyopathy. Alternative splicing results in multiple transcript variants of AIF and a related pseudogene has been identified on chromosome 10.Specifications
| AIFM1 | |
| Monoclonal | |
| Unconjugated | |
| AIFM1 | |
| AIF, CMT2D, CMTX4, COWCK, COXPD6, DFNX5, NADMR, NAMSD, PDCD8 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26926, 9131 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5D2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95831, Q9Z0X1 | |
| AIFM1 | |
| Human recombit protein fragment corresponding to amino acids 134-483 of human AIFM1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction