missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGTR-1 Antibody (S07-1I6), Novus Biologicals™
Rabbit Monoclonal Antibody
421.00 EUR - 645.00 EUR
Specifications
| Antigen | AGTR-1 |
|---|---|
| Clone | S07-1I6 |
| Concentration | 0.3 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686724
|
Novus Biologicals
NBP3-19499-100ul |
100 μg |
645.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18688534
|
Novus Biologicals
NBP3-19499-50ul |
50 μg |
421.00 EUR
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AGTR-1 Monoclonal antibody specifically detects AGTR-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| AGTR-1 | |
| 0.3 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AG2S, AGTR1AAngiotensin II type-1 receptor, Angiostensin Receptor, angiotensin II receptor, type 1, angiotensin receptor 1, angiotensin receptor 1B, AT1AT1AR, AT1B, AT1R, AT2R1A, AT2R1AGTR1B, AT2R1BAT1BR, HAT1R, type-1 angiotensin II receptor, type-1B angiotensin II receptor | |
| A synthetic peptide of human AGTR-1 (Uniprot # P30556) | |
| Primary | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. |
| S07-1I6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 185 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title