missing translation for 'onlineSavingsMsg'
Learn More

AGTPBP1 Monoclonal Antibody (OTI1C8), TrueMAB™, OriGene

Product Code. 17557792
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17557792 30 μL 30µL
17537792 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17557792 Supplier Origene Technologies Supplier No. TA800186S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

AGTPBP1 Monoclonal antibody specifically detects AGTPBP1 in Human samples. It is validated for Western Blot

Specifications

Antigen AGTPBP1
Applications Western Blot
Classification Monoclonal
Clone OTI1C8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene NUB1
Gene Accession No. Q9Y5A7
Gene Alias 1700020N17Rik; 2310001G17Rik; 2900054O13Rik; 4930445M19Rik; 5730402G09Rik; agtpbp1; ATP/GTP binding protein 1; ATP/GTP-binding protein 1; carboxypeptidase-tubulin; CCP1; Cytosolic carboxypeptidase 1; cytosolic carboxypeptidase 1; LOW QUALITY PROTEIN: cytosolic carboxypeptidase 1; Cytosolic carboxypeptidase 1-like protein; DKFZp686M20191; KIAA1035; Nervous system nuclear protein induced by axotomy protein 1; nervous system nuclear protein induced by axotomy protein 1 homolog; nmf243; Nna1; nuclear ATP/GTP-binding protein; pcd; Purkinje cell degeneration; si:ch73-236b16.2; soluble carboxypeptidase; tubulinyl-Tyr carboxypeptidase; tyrosine carboxypeptidase
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-326 of human NUB1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51667
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.