missing translation for 'onlineSavingsMsg'
Learn More

Agmatine sulfate, MedChemExpress

Product Code. 16454556
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
This item is not returnable. View return policy

Product Code. 16454556

Brand: Medchem Express HY101238

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Agmatine sulfate exerts modulatory action at multiple molecular targets, such as neurotransmitter systems, ion channels and nitric oxide synthesis. It is an endogenous agonist at imidazoline receptor and a NO synthase inhibitor.

Agmatine sulfate exerts modulatory action at multiple molecular targets, such as neurotransmitter systems, ion channels and nitric oxide synthesis. It is an endogenous agonist at imidazoline receptor and a NO synthase inhibitor.

Specifications

Chemical Name or Material Agmatine sulfate
Physical Form Solid
CAS 2482-00-0
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H16N4O4S
For Use With (Application) Cancer-programmed cell death
Solubility Information H2O : ≥ 100 mg/mL (438.08 mM) ∣DMSO : 1 mg/mL (4.38 mM; ultrasonic and warming and heat to 80°C)
SMILES NC(NCCCCN)=N.O=S(O)(O)=O
Molecular Weight (g/mol) 228.27
Formula Weight 228.27
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.