missing translation for 'onlineSavingsMsg'
Learn More

AGI-5198, MedChemExpress

Product Code. p-200096165
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16467458

Brand: Medchem Express HY18082

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor with an IC50 of 0.07 μM.

AGI-5198 (IDH-C35) is a potent and selective mutant IDH1R132H inhibitor with an IC50 of 0.07 μM.

Specifications

Chemical Name or Material AGI-5198
CAS 1355326-35-0
Color White
Purity Grade Notes Research
Molecular Formula C27H31FN4O2
Quantity 10 mg
Synonym IDH-C35
Solubility Information DMF : ≥ 50 mg/mL (108.09 mM) ∣DMSO : 20.83 mg/mL (45.03 mM; Need ultrasonic)
SMILES FC1=CC=CC(N(C(C2=CC=CC=C2C)C(NC3CCCCC3)=O)C(CN4C=CN=C4C)=O)=C1
Molecular Weight (g/mol) 462.56
Formula Weight 462.56
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.